Learn More
Trajan Scientific Tubing PEEKSil Purple Medical 2/PK
SDP
Catalog No. 5000494614
Encompass_Preferred
1/32 OD x 0 15 mm ID x 750 mm length PEEKsil tubing purple PK2
Catalog No. 50-004-94614 Supplier Trajan Scientific Supplier No. 14020507500502
1/32 OD x 0 15 mm ID x 750 mm length PEEKsil tubing purple PK2