Learn More
Trajan Scientific Tubing PEEKSil Red Medical 2/PK
SDP
Catalog No. 5000494393
Encompass_Preferred
1/16 OD x 0 1 mm ID x 750 mm length PEEKsil tubing red PK2
Catalog No. 50-004-94393 Supplier Trajan Scientific Supplier No. 14010407500402
1/16 OD x 0 1 mm ID x 750 mm length PEEKsil tubing red PK2