Learn More
Trajan Scientific Tubing PEEKSil Red Medical 2/PK
SDP
Catalog No. 5000493681
Encompass_Preferred
1/32 OD x 0 1 mm ID x 500 mm length PEEKsil tubing red PK2
Catalog No. 50-004-93681 Supplier Trajan Scientific Supplier No. 14020405000402
1/32 OD x 0 1 mm ID x 500 mm length PEEKsil tubing red PK2