Learn More
Trajan Scientific Tubing PEEKSil Yellow Medical 2/PK
SDP
Catalog No. 5000493716
Encompass_Preferred
1/16 OD x 0 175 mm ID x 650 mm length PEEKsil tubing yellow PK2
Catalog No. 50-004-93716 Supplier Trajan Scientific Supplier No. 14010606500602
1/16 OD x 0 175 mm ID x 650 mm length PEEKsil tubing yellow PK2