Learn More
Trajan Scientific Tubing PEEKSil Yellow Medical 2/PK
SDP
Catalog No. 5000493665
Encompass_Preferred
1/16 OD x 0 175 mm ID x 100 mm length PEEKsil tubing yellow PK2
Catalog No. 50-004-93665 Supplier Trajan Scientific Supplier No. 14010601000602
1/16 OD x 0 175 mm ID x 100 mm length PEEKsil tubing yellow PK2