Learn More
Trajan Scientific Tubing PEEKSil Yellow Medical 2/PK
SDP
Catalog No. 5000493714
Encompass_Preferred
1/16 OD x 0 175 mm ID x 400 mm length PEEKsil tubing yellow PK2
Catalog No. 50-004-93714 Supplier Trajan Scientific Supplier No. 14010604000602
1/16 OD x 0 175 mm ID x 400 mm length PEEKsil tubing yellow PK2