Learn More
Trajan Scientific Tubing PEEKSil Yellow Medical 2/PK
SDP
Catalog No. 5000493884
Encompass_Preferred
1/16 OD x 0 175 mm ID x 250 mm length PEEKsil tubing yellow PK2
Catalog No. 50-004-93884 Supplier Trajan Scientific Supplier No. 14010602500602
1/16 OD x 0 175 mm ID x 250 mm length PEEKsil tubing yellow PK2