Learn More
Trajan Scientific Tubing PEEKSil Yellow Medical 2/PK
SDP
Catalog No. 5000493889
Encompass_Preferred
1/16 OD x 0 175 mm ID x 500 mm length PEEKsil tubing yellow PK2
Catalog No. 50-004-93889 Supplier Trajan Scientific Supplier No. 14010605000602
1/16 OD x 0 175 mm ID x 500 mm length PEEKsil tubing yellow PK2