Learn More
IDEX Health & Science Upchurch TUBING PFA .030INX1/16INX50'
Catalog No. NC1919827
Encompass_Preferred
Idex PFA Tubing, Diameter Inner: 0.03 in., 0.76 mm, Diameter Outer: 0.0625 in., 1.58 mm, Overall Length: 50 ft., 15.24 m, Pressure: 1000 psi, Natural
Catalog No. NC1919827 Supplier IDEX Health & Science Upchurch Supplier No. 1514L
Idex PFA Tubing, Diameter Inner: 0.03 in., 0.76 mm, Diameter Outer: 0.0625 in., 1.58 mm, Overall Length: 50 ft., 15.24 m, Pressure: 1000 psi, Natural