Learn More
Microsolv Technology Corporation Tubing, PFA, 0.040" ID, 1/16" OD, Low Pressure. 5 Meter Roll. 1 EA.
Catalog No. 502271517
Encompass_Preferred
This MicroSolv PFA ( PerFlourAlkoxy ) low pressure tubing is very similar to Teflon but is less permeable, more transparent and performs better in high temperature and pressure. Higher temperature than FEP are possible. This tubing is used in all low-pressure, laboratory applications including before and after an HPLC Pump such as with Mobile Phase Delivery and Solvent Waste Systems. Also known as Teflon FEP.Do not use this tubing with heat over 260⁰C. Internal and Outer Dimensional Tolerance: +/- 0.10mm Color: Natural PFA, Transparent, slight light blue tint ID: 0.040" ( 1.0 mm ) Pressure rating: 800 psi.
This item is not returnable. View return policy
Catalog No. 50-227-1517 Supplier Microsolv Technology Corporation Supplier No. 4831040
This MicroSolv PFA ( PerFlourAlkoxy ) low pressure tubing is very similar to Teflon but is less permeable, more transparent and performs better in high temperature and pressure. Higher temperature than FEP are possible. This tubing is used in all low-pressure, laboratory applications including before and after an HPLC Pump such as with Mobile Phase Delivery and Solvent Waste Systems. Also known as Teflon FEP.Do not use this tubing with heat over 260⁰C. Internal and Outer Dimensional Tolerance: +/- 0.10mm Color: Natural PFA, Transparent, slight light blue tint ID: 0.040" ( 1.0 mm ) Pressure rating: 800 psi.