Learn More
IDEX Health & Science Upchurch TUBING PFA 1/16X1/8X50'
Catalog No. NC1919824
Encompass_Preferred
Idex PFA Tubing, Diameter Inner: 0.062 in., 1.57 mm, Diameter Outer: 0.125 in., 3.17 mm, Overall Length: 50 ft., 15.24 m, Pressure: 500 psi, Natural
Catalog No. NC1919824 Supplier IDEX Health & Science Upchurch Supplier No. 1509L
Idex PFA Tubing, Diameter Inner: 0.062 in., 1.57 mm, Diameter Outer: 0.125 in., 3.17 mm, Overall Length: 50 ft., 15.24 m, Pressure: 500 psi, Natural