Learn More
Waters Corp Tubing, PFA, 1/4 in. OD, 5/32 in. ID, 25 ft.
Catalog No. 502391193 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
PFA tubing is commonly used in laboratories and industrial settings due to its excellent chemical resistance and durability, providing reliable fluid transfer for a variety of applications.
  • Constructed from PFA (Perfluoroalkoxy) material.
  • Outer diameter of 1/4 in.
  • Inner diameter of 5/32 in.
  • Supplied in a 25 ft. length.
  • Offers high chemical resistance for diverse fluid compatibility.
  • Designed for critical fluid transfer and analytical applications.
Catalog No. 50-239-1193 Supplier Waters Corp Supplier No. 700002492
PFA tubing is commonly used in laboratories and industrial settings due to its excellent chemical resistance and durability, providing reliable fluid transfer for a variety of applications.
  • Constructed from PFA (Perfluoroalkoxy) material.
  • Outer diameter of 1/4 in.
  • Inner diameter of 5/32 in.
  • Supplied in a 25 ft. length.
  • Offers high chemical resistance for diverse fluid compatibility.
  • Designed for critical fluid transfer and analytical applications.