Learn More
Microsolv Technology Corporation Tubing Pfa 1/8 X 1/16Th 5M
Catalog No. 5001451606
Encompass_Preferred
Tubing, PFA, 1/16 ID, 1/8 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51606 Supplier Microsolv Technology Corporation Supplier No. 4833565
Tubing, PFA, 1/16 ID, 1/8 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.