Learn More
AVANTOR MASTERFLEX (VWR LLC) Dow Corning Pharma-50 Tubing, 3/16"" x 3/8"", 50 ft Pack
Catalog No. NC1315519
Encompass
Applications: Pharmaceutical, biotechnology , and ultra-pure fluid processing. Characteristics: 50 durometer, translucent platinum-cured silicone tubing with a very low extractable profile; it contains no phthalates, plasticizers, latex, or other organic additives. Pharma tubing remains flexible and resilient for long service life. Dow Corning maintains complete traceability from silica to silicone tubing. Certifications: Elastomer exceeds test guidelines for USP Class VI Plastics, and the tubing itself is FDA compliant, and meets 3A Sanitary Standards and European Pharmacopoeia (EP) requirements. Produced under Bulk Pharmaceutical (BP) and current GMP principles. Temperature range: ¿60 to 460¿F (¿51 to 232¿C) Sterilize: Autoclave, ethylene oxide, or gamma irradiation.
Catalog No. NC1315519 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9610502
Applications: Pharmaceutical, biotechnology , and ultra-pure fluid processing. Characteristics: 50 durometer, translucent platinum-cured silicone tubing with a very low extractable profile; it contains no phthalates, plasticizers, latex, or other organic additives. Pharma tubing remains flexible and resilient for long service life. Dow Corning maintains complete traceability from silica to silicone tubing. Certifications: Elastomer exceeds test guidelines for USP Class VI Plastics, and the tubing itself is FDA compliant, and meets 3A Sanitary Standards and European Pharmacopoeia (EP) requirements. Produced under Bulk Pharmaceutical (BP) and current GMP principles. Temperature range: ¿60 to 460¿F (¿51 to 232¿C) Sterilize: Autoclave, ethylene oxide, or gamma irradiation.