Learn More
Waters Corp Tubing, Pharmed, 1/4 in. OD, 1/8 in. ID, 5 ft.
Catalog No. 502392989 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Pharmed tubing is designed for various laboratory applications. It features precise dimensions suitable for connecting different components in a system.
  • Outer diameter: 1/4 in.
  • Inner diameter: 1/8 in.
  • Length: 5 ft.
  • Constructed from Pharmed material for compatibility.
Catalog No. 50-239-2989 Supplier Waters Corp Supplier No. 700002181
This Pharmed tubing is designed for various laboratory applications. It features precise dimensions suitable for connecting different components in a system.
  • Outer diameter: 1/4 in.
  • Inner diameter: 1/8 in.
  • Length: 5 ft.
  • Constructed from Pharmed material for compatibility.