Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S 2-Stop High-Performance Precision Pump Tubing, PharMed BPT, L/S 15; Pack of 4
Catalog No. NC1041580
Encompass
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige For use with Masterflex multichannel pump heads.
Catalog No. NC1041580 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9611415/DEL
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige For use with Masterflex multichannel pump heads.