Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, PharMed BPT, 3/16"" ID x 5/16"" OD; 25 ft
Catalog No. 501961051
Encompass
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165¿F (-46 to 73¿C). Can be sterilized with ethylene oxide or autoclave.
Catalog No. 50-196-1051 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9688008
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165¿F (-46 to 73¿C). Can be sterilized with ethylene oxide or autoclave.