Learn More
Thermo Scientific Tubing 0.010" ID 5 ft PK 13.46
Supplier:  Thermo Scientific 052306
Encompass
0.025 inch outer diameter 5 foot long polypropylene capillary tubing with 0.010 inch inner diameter for fluid transfer and analytical instrumentation applications requiring precise small volume control and chemical resistance to solvents.
Catalog No. 50-134-5090