Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Platinum-Cured Silicone, 1/4"" ID x 3/8"" OD; 300 ft
Catalog No. 501961053
Encompass
Reduce unpacking time, save money on shipping and ensure that you always have enough tubing on hand with bulk tubing. Platinum-cured silicone spools are sold in 300 ft or 500 ft spools.
Catalog No. 50-196-1053 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9890608
Reduce unpacking time, save money on shipping and ensure that you always have enough tubing on hand with bulk tubing. Platinum-cured silicone spools are sold in 300 ft or 500 ft spools.