Learn More
Microsolv Technology Corporation Tubing Ptfe 1/16 X .040 5M
Catalog No. 5001451624
Encompass_Preferred
Tubing, PTFE, 0.040 ID, 1/16 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51624 Supplier Microsolv Technology Corporation Supplier No. 4921040
Tubing, PTFE, 0.040 ID, 1/16 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.