Learn More
Cole-Parmer Diba Omnifit Tubing, PTFE, 1/16" (1.6 mm) OD x 0.8 mm ID; 20 m/pk
Catalog No. NC2101626 Shop All Cole-Parmer Products
Encompass_Preferred
Use chemically inert PTFE tubing with virtually all chemicals, solvents, and corrosive materials, even at elevated temperatures. Autoclave tubing or sterilize in-line by steam or chemical methods. Select from 1/16", 1/8", or 3/16" OD tubing with a range of inside diameters.
This item is not returnable. View return policy
Catalog No. NC2101626 Supplier Cole-Parmer Supplier No. 2194272
Use chemically inert PTFE tubing with virtually all chemicals, solvents, and corrosive materials, even at elevated temperatures. Autoclave tubing or sterilize in-line by steam or chemical methods. Select from 1/16", 1/8", or 3/16" OD tubing with a range of inside diameters.