Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, PTFE, 1/4"" ID x 3/8"" OD; 25 ft
Catalog No. 50634373
Encompass
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Good for high-purity work. PTFE tubing withstands extreme temperatures. Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent. Certification: Resin is FDA-compliant (21 CFR 177.1550) Temperature range: -400 to 500¿F (-240 to 260¿C) Sterilize: Autoclave, ethylene oxide, or dry heat.
Catalog No. 50-634-373 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0660535
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Good for high-purity work. PTFE tubing withstands extreme temperatures. Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent. Certification: Resin is FDA-compliant (21 CFR 177.1550) Temperature range: -400 to 500¿F (-240 to 260¿C) Sterilize: Autoclave, ethylene oxide, or dry heat.