Learn More
Microsolv Technology Corporation Tubing, PTFE, 1.70" ID, 2mm OD, Low Pressure. 5 Meter Roll. 1 EA.
Catalog No. 502271527
Encompass_Preferred
This MicroSolv, Thin Walled, PTFE ( Polytetrafluoroethylene ) Tubing is extruded from virgin PTFE, manufactured exclusively by DuPont. We do not add any additives to the original PTFE material, except for particles for color coding. This tubing is used widely in all low-pressure, laboratory applications including before and after an HPLC Pump such as with Mobile Phase Delivery and Solvent Waste Systems. Since PTFE tubing contains "micro-pores" in the tubing wall, which allows a quick air exchange, consideration for for use with degassed and oxygen-sensitive solvents is needed. Internal and Outer Dimensional Tolerance: +/- 0.20mm Color: Natural PTFE, Translucent ID: 1.70mmPressure rating: 150 psi.
This item is not returnable. View return policy
Catalog No. 50-227-1527 Supplier Microsolv Technology Corporation Supplier No. 4921070
This MicroSolv, Thin Walled, PTFE ( Polytetrafluoroethylene ) Tubing is extruded from virgin PTFE, manufactured exclusively by DuPont. We do not add any additives to the original PTFE material, except for particles for color coding. This tubing is used widely in all low-pressure, laboratory applications including before and after an HPLC Pump such as with Mobile Phase Delivery and Solvent Waste Systems. Since PTFE tubing contains "micro-pores" in the tubing wall, which allows a quick air exchange, consideration for for use with degassed and oxygen-sensitive solvents is needed. Internal and Outer Dimensional Tolerance: +/- 0.20mm Color: Natural PTFE, Translucent ID: 1.70mmPressure rating: 150 psi.