Learn More
Microsolv Technology Corporation Tubing Ptfe 1/8 X 1/100 5M
Catalog No. 5001451625
Encompass_Preferred
Tubing, PTFE, 2.4mm ID, 1/8 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51625 Supplier Microsolv Technology Corporation Supplier No. 49225100
Tubing, PTFE, 2.4mm ID, 1/8 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.