Learn More
Microsolv Technology Corporation Tubing Holder, Clamp, for 1/8" or 1/16" OD Tubing. Blue. 10/PK.
Catalog No. 502271446
Encompass_Preferred
This Tubing Holder will hold 1/16" or 1/8" OD Tubing in place on a beaker, bottle or any thin walled vessel. The spring loaded holder is excellent while draining or siphoning or filling with tubing. Equipped with 2 holes, one hole accommodates 1/16" and the other accommodates 1/8" tubing.
This item is not returnable. View return policy
Catalog No. 50-227-1446 Supplier Microsolv Technology Corporation Supplier No. 4660001
This Tubing Holder will hold 1/16" or 1/8" OD Tubing in place on a beaker, bottle or any thin walled vessel. The spring loaded holder is excellent while draining or siphoning or filling with tubing. Equipped with 2 holes, one hole accommodates 1/16" and the other accommodates 1/8" tubing.