Learn More
Chemglass Life Sciences 3/16” ID X 1/4” OD X 2-1/2” Long PTFE Delivery Stem
SDP
Catalog No. 50872615
Encompass_Preferred
Pack of 12 PTFE delivery stems for use with the CG1049C adapter series. Design incorporates a drip tube sealed within thread to prevent breakage and are supplied with a 2-1/2¿ PTFE delivery stem. Hose connection is 10mm OD at the largest serration. 3/16¿ ID X 1/4¿ OD X 2-1/2¿ Long PTFE Delivery Stem.
Catalog No. 50-872-615 Supplier Chemglass Life Sciences Supplier No. CG1049C50
Pack of 12 PTFE delivery stems for use with the CG1049C adapter series. Design incorporates a drip tube sealed within thread to prevent breakage and are supplied with a 2-1/2¿ PTFE delivery stem. Hose connection is 10mm OD at the largest serration. 3/16¿ ID X 1/4¿ OD X 2-1/2¿ Long PTFE Delivery Stem.