Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Metric PTFE, 7.98 mm ID x 10.0 mm OD; 25 ft
Catalog No. 50634393
Encompass
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Good for high-purity work. PTFE tubing withstands extreme temperatures. Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent. Resin certification: Resin is FDA-compliant (21 CFR 177.1550); meets USP 23 Class VI requirements. Tubing is manufactured according to GMP. Temperature range: -400 to 500¿F (-240 to 260¿C) Sterilize: With autoclave, ethylene oxide, or use dry heat.
Catalog No. 50-634-393 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0660556
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Good for high-purity work. PTFE tubing withstands extreme temperatures. Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent. Resin certification: Resin is FDA-compliant (21 CFR 177.1550); meets USP 23 Class VI requirements. Tubing is manufactured according to GMP. Temperature range: -400 to 500¿F (-240 to 260¿C) Sterilize: With autoclave, ethylene oxide, or use dry heat.