Learn More
Waters Corp Waters Corporation Tubing, PTFE, Thin Wall, 2 in.
Catalog No. 502039638 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement tubing is designed for connecting solvent filter assemblies and solvent inlet lines on various sample and solvent managers, separation modules, and HPLC pumps.
  • Replacement tubing for solvent filter assemblies and solvent inlet lines.
  • Compatible with a range of analytical systems, including various sample managers, separation modules, and HPLC pumps.
  • Suitable for carbohydrate applications.
  • Applicable to HPLC, UPLC, and purification system types.
Catalog No. 50-203-9638 Supplier Waters Corp Supplier No. 700009143
This replacement tubing is designed for connecting solvent filter assemblies and solvent inlet lines on various sample and solvent managers, separation modules, and HPLC pumps.
  • Replacement tubing for solvent filter assemblies and solvent inlet lines.
  • Compatible with a range of analytical systems, including various sample managers, separation modules, and HPLC pumps.
  • Suitable for carbohydrate applications.
  • Applicable to HPLC, UPLC, and purification system types.