Learn More
Instech Polyurethane tubing, .017x.037in. Thick walled to minimize kinking or puncture. Makes a good connection with 25ga couplers and swivels. 30m (98ft) spool, nonsterile. For external use only; not for implantation.
SDP
Catalog No. 5081250
Encompass
The Instech VAHBPU‑T25 is a polyurethane external infusion tubing designed for dependable connections with 25ga swivels and couplers. Its thick‑walled .017x.037in construction resists kinking and puncture while providing a secure friction fit for tether and infusion setups. Supplied nonsterile on a 30m (98ft) spool, it is intended for external infusion use only and not for implantation.
Features
  • Polyurethane tubing with .017x.037in dimensions
  • Thick‑walled to minimize kinking and puncture
  • Reliable fit with 25ga couplers and swivels
  • 30m (98ft) nonsterile spool
  • For external infusion use only; not implantable
  • Used in VAH and VAB tether assemblies
Catalog No. 50-812-50 Supplier Instech Supplier No. VAHBPUT25
The Instech VAHBPU‑T25 is a polyurethane external infusion tubing designed for dependable connections with 25ga swivels and couplers. Its thick‑walled .017x.037in construction resists kinking and puncture while providing a secure friction fit for tether and infusion setups. Supplied nonsterile on a 30m (98ft) spool, it is intended for external infusion use only and not for implantation.
Features
  • Polyurethane tubing with .017x.037in dimensions
  • Thick‑walled to minimize kinking and puncture
  • Reliable fit with 25ga couplers and swivels
  • 30m (98ft) nonsterile spool
  • For external infusion use only; not implantable
  • Used in VAH and VAB tether assemblies