Learn More
Instech Polyurethane tubing, .025x.055in. Thick walled to minimize kinking or puncture. Makes a good connection with 22ga couplers and swivels. 30m (98ft) spool, nonsterile. For external use only; not for implantation.
SDP
Catalog No. 5081249
Encompass
The Instech VAHBPU‑T22 is a thick‑walled polyurethane external infusion tubing sized at .025x.055in (0.6x1.4mm), designed to provide a reliable friction fit with 22ga swivels and couplers for use in VAH and VAB tether assemblies. Its durable construction minimizes kinking, puncture, and accidental disconnection, and it is supplied non-sterile on a 30m (98ft) spool for external infusion applications only.
Features
  • Polyurethane external infusion tubing
  • .025x.055in (0.6x1.4mm) dimensions
  • 30m (98ft) non-sterile spool
  • Thick-walled tubing resists kinking and puncture
  • Reliable friction fit with 22ga swivels and couplers
  • Used in VAH and VAB tether assemblies
  • For external infusion use only; not implantable
Catalog No. 50-812-49 Supplier Instech Supplier No. VAHBPUT22
 Restricted Item
Add to Cart
Add to Cart
The Instech VAHBPU‑T22 is a thick‑walled polyurethane external infusion tubing sized at .025x.055in (0.6x1.4mm), designed to provide a reliable friction fit with 22ga swivels and couplers for use in VAH and VAB tether assemblies. Its durable construction minimizes kinking, puncture, and accidental disconnection, and it is supplied non-sterile on a 30m (98ft) spool for external infusion applications only.
Features
  • Polyurethane external infusion tubing
  • .025x.055in (0.6x1.4mm) dimensions
  • 30m (98ft) non-sterile spool
  • Thick-walled tubing resists kinking and puncture
  • Reliable friction fit with 22ga swivels and couplers
  • Used in VAH and VAB tether assemblies
  • For external infusion use only; not implantable