Learn More
Instech Polyurethane tubing, medical grade, 1Fr. .007x.014in (0.18x0.36mm). 10m (33ft) spool, non-sterile.
SDP
Catalog No. 50890054
Encompass
The Instech BTPU‑014 is a 1Fr polyurethane intravascular tubing with a .007x.014in (0.18x0.36mm) profile, offering excellent flexibility, biocompatibility, and friction‑fit performance for small‑animal catheter applications. Supplied non‑sterile on a 10m (33ft) spool, it is commonly used in fabrication of mouse catheters and other micro‑bore implanted lines where stretch, durability, and precise flow characteristics are required.
Features
  • Polyurethane intravascular tubing for small‑animal catheterization
  • .007x.014in (0.18x0.36mm) dimensions; 1Fr size
  • 10m (33ft) non‑sterile spool
  • Good biocompatibility for implantation
  • Stretchy material for secure friction‑fit connections
  • Suitable for custom catheter fabrication in research workflows
Catalog No. 50-890-054 Supplier Instech Supplier No. BTPU014
The Instech BTPU‑014 is a 1Fr polyurethane intravascular tubing with a .007x.014in (0.18x0.36mm) profile, offering excellent flexibility, biocompatibility, and friction‑fit performance for small‑animal catheter applications. Supplied non‑sterile on a 10m (33ft) spool, it is commonly used in fabrication of mouse catheters and other micro‑bore implanted lines where stretch, durability, and precise flow characteristics are required.
Features
  • Polyurethane intravascular tubing for small‑animal catheterization
  • .007x.014in (0.18x0.36mm) dimensions; 1Fr size
  • 10m (33ft) non‑sterile spool
  • Good biocompatibility for implantation
  • Stretchy material for secure friction‑fit connections
  • Suitable for custom catheter fabrication in research workflows