Learn More
Instech Polyurethane tubing, medical grade, 2Fr. .017x.027in (0.3x0.6mm). 30m (98ft) spool, non-sterile.
SDP
Catalog No. 50890055
Encompass
The Instech BTPU‑027 is a 2Fr polyurethane intravascular tubing with a .017x.027in (0.43x0.69mm) profile, providing a flexible, biocompatible, kink‑resistant flow path ideal for rat or mouse catheter fabrication. Sized to fit 25ga connectors, it offers excellent friction‑fit performance and durability for implanted catheter use. Supplied non‑sterile on a 30m (98ft) spool, it is widely used in research workflows requiring reliable small‑bore polyurethane tubing.
Features
  • Polyurethane intravascular tubing for small‑animal catheterization
  • .017x.027in (0.43x0.69mm) dimensions; 2Fr size
  • 30m (98ft) non‑sterile spool
  • Fits 25ga connectors, swivels, and couplers
  • Good biocompatibility and kink resistance
  • Ideal for custom catheter fabrication in research applications
Catalog No. 50-890-055 Supplier Instech Supplier No. BTPU027
The Instech BTPU‑027 is a 2Fr polyurethane intravascular tubing with a .017x.027in (0.43x0.69mm) profile, providing a flexible, biocompatible, kink‑resistant flow path ideal for rat or mouse catheter fabrication. Sized to fit 25ga connectors, it offers excellent friction‑fit performance and durability for implanted catheter use. Supplied non‑sterile on a 30m (98ft) spool, it is widely used in research workflows requiring reliable small‑bore polyurethane tubing.
Features
  • Polyurethane intravascular tubing for small‑animal catheterization
  • .017x.027in (0.43x0.69mm) dimensions; 2Fr size
  • 30m (98ft) non‑sterile spool
  • Fits 25ga connectors, swivels, and couplers
  • Good biocompatibility and kink resistance
  • Ideal for custom catheter fabrication in research applications