Learn More
Instech Polyurethane tubing, medical grade, 32GA/0.8Fr. .005x.010in (0.13x0.25mm). 10m (33ft) spool, non-sterile.
SDP
Catalog No. 50890053
Encompass
The Instech BTPU‑010 is a 32ga polyurethane intravascular tubing with a .005x.010in (0.13x0.25mm) profile, providing a flexible and biocompatible micro‑bore flow path suitable for mouse, intrathecal, or other ultra‑small catheter applications. Supplied non‑sterile on a 10m (33ft) spool, it offers excellent friction‑fit performance on micro‑connectors and is commonly used for custom catheter fabrication in research settings.
Features
  • Polyurethane intravascular tubing for micro‑catheter applications
  • .005x.010in (0.13x0.25mm) dimensions; approx. 32ga / 0.8Fr size
  • 10m (33ft) non‑sterile spool
  • Good biocompatibility for implantation
  • Stretchy material for secure friction‑fit connections
  • Ideal for microscale infusion and catheter fabrication
Catalog No. 50-890-053 Supplier Instech Supplier No. BTPU010
The Instech BTPU‑010 is a 32ga polyurethane intravascular tubing with a .005x.010in (0.13x0.25mm) profile, providing a flexible and biocompatible micro‑bore flow path suitable for mouse, intrathecal, or other ultra‑small catheter applications. Supplied non‑sterile on a 10m (33ft) spool, it offers excellent friction‑fit performance on micro‑connectors and is commonly used for custom catheter fabrication in research settings.
Features
  • Polyurethane intravascular tubing for micro‑catheter applications
  • .005x.010in (0.13x0.25mm) dimensions; approx. 32ga / 0.8Fr size
  • 10m (33ft) non‑sterile spool
  • Good biocompatibility for implantation
  • Stretchy material for secure friction‑fit connections
  • Ideal for microscale infusion and catheter fabrication