Learn More
Instech Polyurethane tubing, medical grade, 3Fr. .025x.040in (0.6x1.0mm). 30m (98ft) spool, non-sterile.
SDP
Catalog No. 50890056
Encompass
The Instech BTPU‑040 is a 3Fr polyurethane intravascular tubing with a .025x.040in (0.64x1.01mm) profile, offering excellent flexibility, biocompatibility, and kink resistance for implanted catheter applications. Designed to fit 22ga connectors, it provides a secure friction‑fit interface ideal for rat catheterization and other small‑animal infusion or sampling procedures. Supplied non‑sterile on a 30m (98ft) spool, it is widely used for fabricating durable laboratory research catheters.
Features
  • Polyurethane intravascular tubing for small‑animal catheterization
  • .025x.040in (0.64x1.01mm) dimensions; 3Fr size
  • 30m (98ft) non‑sterile spool
  • Fits 22ga connectors, swivels, and couplers
  • Good biocompatibility and kink resistance
  • Stretchy material for secure friction‑fit connections
Catalog No. 50-890-056 Supplier Instech Supplier No. BTPU040
The Instech BTPU‑040 is a 3Fr polyurethane intravascular tubing with a .025x.040in (0.64x1.01mm) profile, offering excellent flexibility, biocompatibility, and kink resistance for implanted catheter applications. Designed to fit 22ga connectors, it provides a secure friction‑fit interface ideal for rat catheterization and other small‑animal infusion or sampling procedures. Supplied non‑sterile on a 30m (98ft) spool, it is widely used for fabricating durable laboratory research catheters.
Features
  • Polyurethane intravascular tubing for small‑animal catheterization
  • .025x.040in (0.64x1.01mm) dimensions; 3Fr size
  • 30m (98ft) non‑sterile spool
  • Fits 22ga connectors, swivels, and couplers
  • Good biocompatibility and kink resistance
  • Stretchy material for secure friction‑fit connections