Learn More
Hach Company Tubing, PVC 0.025 in (0.635 mm) I.D., Orange/White - Pump tubing, 1/pack
Catalog No. NC3371166 Shop All Hach Company Products
Encompass
PVC pump tubing with a 0.025 in (0.635 mm) internal diameter supplied as a single-piece pack. It is intended for small-diameter liquid transfer and replacement in compatible pump systems.
  • Flexible PVC construction for reliable liquid transfer.
  • 0.025 in (0.635 mm) internal diameter for low-flow applications.
  • Orange/white color coding for easy identification.
  • Supplied as one piece per pack for replacements or repairs.
  • Suitable for compatible peristaltic and metering pump systems.
Catalog No. NC3371166 Supplier Hach Company Supplier No. 53406L
PVC pump tubing with a 0.025 in (0.635 mm) internal diameter supplied as a single-piece pack. It is intended for small-diameter liquid transfer and replacement in compatible pump systems.
  • Flexible PVC construction for reliable liquid transfer.
  • 0.025 in (0.635 mm) internal diameter for low-flow applications.
  • Orange/white color coding for easy identification.
  • Supplied as one piece per pack for replacements or repairs.
  • Suitable for compatible peristaltic and metering pump systems.