Learn More
Chemglass Life Sciences Tubing, Quartz, 10mm ID x 14mm OD, 4ft Length
SDP
Catalog No. 50872642
Encompass_Preferred
Tubing, Quartz, 10mm ID x 14mm OD, 4ft Length. General Electric Type 214 grade clear fused quartz tubing. Tubing is stocked in 4ft lengths. PLEASE NOTE: PRICE LISTED IS PER LENGTH OF 4ft TUBING.
Catalog No. 50-872-642 Supplier Chemglass Life Sciences Supplier No. CGQ0800T25
Tubing, Quartz, 10mm ID x 14mm OD, 4ft Length. General Electric Type 214 grade clear fused quartz tubing. Tubing is stocked in 4ft lengths. PLEASE NOTE: PRICE LISTED IS PER LENGTH OF 4ft TUBING.