Learn More
Chemglass Life Sciences Tubing, Quartz, 19mm ID x 22mm OD, 4ft Length
SDP
Catalog No. 501532067
Encompass_Preferred
Tubing, Quartz, 19mm ID x 22mm OD, 4ft Length. General Electric Type 214 grade clear fused quartz tubing. Tubing is stocked in 4ft lengths.
Catalog No. 50-153-2067 Supplier Chemglass Life Sciences Supplier No. CGQ0800T59
Tubing, Quartz, 19mm ID x 22mm OD, 4ft Length. General Electric Type 214 grade clear fused quartz tubing. Tubing is stocked in 4ft lengths.