Learn More
Chemglass Life Sciences Tubing, Quartz, 24mm ID x 28mm OD, 4ft Length
SDP
Catalog No. 501532071
Encompass_Preferred
Tubing, Quartz, 24mm ID x 28mm OD, 4ft Length. General Electric Type 214 grade clear fused quartz tubing. Tubing is stocked in 4ft lengths.
Catalog No. 50-153-2071 Supplier Chemglass Life Sciences Supplier No. CGQ0800T74
Tubing, Quartz, 24mm ID x 28mm OD, 4ft Length. General Electric Type 214 grade clear fused quartz tubing. Tubing is stocked in 4ft lengths.