Learn More
Chemglass Life Sciences Tubing, Quartz, 28mm ID x 32mm OD, 4ft Length
SDP
Supplier:  Chemglass Life Sciences CGQ0800T91
Encompass_Preferred
Tubing, Quartz, 28mm ID x 32mm OD, 4ft Length. General Electric Type 214 grade clear fused quartz tubing. Tubing is stocked in 4ft lengths.
Catalog No. 50-153-2074