Learn More
Chemglass Life Sciences Tubing, Quartz, 2mm ID x 4mm OD, 4ft Length
SDP
Catalog No. 50872628
Encompass_Preferred
Tubing, Quartz, 2mm ID x 4mm OD, 4ft Length. General Electric Type 214 grade clear fused quartz tubing. Tubing is stocked in 4ft lengths. PLEASE NOTE: PRICE LISTED IS PER LENGTH OF 4ft TUBING.
Catalog No. 50-872-628 Supplier Chemglass Life Sciences Supplier No. CGQ0800T03
Tubing, Quartz, 2mm ID x 4mm OD, 4ft Length. General Electric Type 214 grade clear fused quartz tubing. Tubing is stocked in 4ft lengths. PLEASE NOTE: PRICE LISTED IS PER LENGTH OF 4ft TUBING.