Learn More
Chemglass Life Sciences Tubing, Quartz, 6mm ID x 10mm OD, 4ft Length
SDP
Catalog No. 50872636
Encompass_Preferred
Tubing, Quartz, 6mm ID x 10mm OD, 4ft Length. General Electric Type 214 grade clear fused quartz tubing. Tubing is stocked in 4ft lengths. PLEASE NOTE: PRICE LISTED IS PER LENGTH OF 4ft TUBING.
Catalog No. 50-872-636 Supplier Chemglass Life Sciences Supplier No. CGQ0800T11
Tubing, Quartz, 6mm ID x 10mm OD, 4ft Length. General Electric Type 214 grade clear fused quartz tubing. Tubing is stocked in 4ft lengths. PLEASE NOTE: PRICE LISTED IS PER LENGTH OF 4ft TUBING.