Learn More
Med Vet International Medline Suction Tubing, 0.188 Inch Inner Diameter, 2 Yard Length, Sterile, DYND50216, 50/Case
Catalog No. 503045067
Encompass_Preferred
6 foot-long suction connector tubing is made of polyvinyl chloride (PVC) and has an inner diameter of 0.188 inch Smooth outer tube surface
Catalog No. 50-304-5067 Supplier Med Vet International Supplier No. 277450CS
6 foot-long suction connector tubing is made of polyvinyl chloride (PVC) and has an inner diameter of 0.188 inch Smooth outer tube surface