Learn More
Instech Complete tubing set for Instech ABS2, 020 silicone, no low loss. For automated blood sampling from rats.
SDP
Catalog No. 5080992
Encompass
The Instech ABS2/TS020NL is a complete tubing set designed for the ABS2™ Automated Blood Sampler. Built from 020-size silicone tubing in a no–low-loss configuration, it supports accurate automated blood sampling by maintaining reliable fluid flow and minimizing dead volume. This tubing set is optimized for rat studies and integrates directly with ABS2 peristaltic pump pathways for consistent sampling performance.
Features
  • Tubing Type: 020 silicone
  • Configuration: No low loss
  • Application: Automated blood sampling from rats
  • Compatibility: ABS2 Automated Blood Sampler
  • Function: Maintains steady fluid flow for sampling and IV fluid return
  • Quantity: 1 complete tubing set
Catalog No. 50-809-92 Supplier Instech Supplier No. ABS2/TS020NL
The Instech ABS2/TS020NL is a complete tubing set designed for the ABS2™ Automated Blood Sampler. Built from 020-size silicone tubing in a no–low-loss configuration, it supports accurate automated blood sampling by maintaining reliable fluid flow and minimizing dead volume. This tubing set is optimized for rat studies and integrates directly with ABS2 peristaltic pump pathways for consistent sampling performance.
Features
  • Tubing Type: 020 silicone
  • Configuration: No low loss
  • Application: Automated blood sampling from rats
  • Compatibility: ABS2 Automated Blood Sampler
  • Function: Maintains steady fluid flow for sampling and IV fluid return
  • Quantity: 1 complete tubing set