Learn More
Instech Complete large animal PET tubing set for Instech ABS210P
SDP
Catalog No. 501004879
Encompass
The Instech ABS2/TS031P is a complete large-animal PET tubing set designed for the ABS2 and ABS210P Automated Blood Sampler systems. Built for high‑speed automated blood sampling in combination with PET imaging, this tubing set provides reliable fluid handling and maintains proper flow characteristics required for large‑animal studies.
Features
  • Tubing Type: Large-animal PET tubing set
  • Application: High-speed automated blood sampling with PET imaging
  • Compatibility: ABS2 and ABS210P automated blood sampling systems
  • Function: Supports precise sampling and IV fluid return in large animals
  • Configuration: Complete tubing set for immediate system integration
  • Quantity: 1 tubing set
Catalog No. 50-100-4879 Supplier Instech Supplier No. ABS2/TS031P
 Restricted Item
Add to Cart
Add to Cart
The Instech ABS2/TS031P is a complete large-animal PET tubing set designed for the ABS2 and ABS210P Automated Blood Sampler systems. Built for high‑speed automated blood sampling in combination with PET imaging, this tubing set provides reliable fluid handling and maintains proper flow characteristics required for large‑animal studies.
Features
  • Tubing Type: Large-animal PET tubing set
  • Application: High-speed automated blood sampling with PET imaging
  • Compatibility: ABS2 and ABS210P automated blood sampling systems
  • Function: Supports precise sampling and IV fluid return in large animals
  • Configuration: Complete tubing set for immediate system integration
  • Quantity: 1 tubing set