Learn More
Integra Biosciences Corp tubing set for the blood agar
SDP
Catalog No. 5000882028
Encompass
Tubing set for the preparation of blood agar (suitable to add 3 to 10% blood in connection with the INTEGRA peristaltic pump)Comprising Tubing (6 x 9 mm)Filling nozzle 103 032Borosilicate glass T-piece 103505Suction needle stainless steel 179 147Tubing (2 x 5 mm) 171022
Catalog No. 50-008-82028 Supplier Integra Biosciences Corp Supplier No. 103040
Tubing set for the preparation of blood agar (suitable to add 3 to 10% blood in connection with the INTEGRA peristaltic pump)Comprising Tubing (6 x 9 mm)Filling nozzle 103 032Borosilicate glass T-piece 103505Suction needle stainless steel 179 147Tubing (2 x 5 mm) 171022