Learn More
Instech Replacement waste tubing segments and vials for Instech ABS2 automated blood sampler
SDP
Catalog No. NC0079210
Encompass
The Instech ABS2/TS062-W5 is a sterile tubing set for the ABS2 automated blood sampler, providing the fluid path needed for drawing blood, returning IV fluid, and routing samples to the refrigerated collector. Designed for use across mouse, rat, and large-animal ABS2 configurations, this set supports accurate, low-loss sampling and maintains closed-system operation for pharmacokinetic studies.
Features
  • Sterile tubing set for ABS2 sampler
  • Supports blood draw and IV return paths
  • Compatible with multi-species ABS2 setups
  • Low-loss routing for undiluted samples
  • Closed-system design for PK workflows
Catalog No. NC0079210 Supplier Instech Supplier No. ABS2/TS062W5
The Instech ABS2/TS062-W5 is a sterile tubing set for the ABS2 automated blood sampler, providing the fluid path needed for drawing blood, returning IV fluid, and routing samples to the refrigerated collector. Designed for use across mouse, rat, and large-animal ABS2 configurations, this set supports accurate, low-loss sampling and maintains closed-system operation for pharmacokinetic studies.
Features
  • Sterile tubing set for ABS2 sampler
  • Supports blood draw and IV return paths
  • Compatible with multi-species ABS2 setups
  • Low-loss routing for undiluted samples
  • Closed-system design for PK workflows