Learn More
Helix Medical Inc TUBING SIL .031 X .157 25FT
Catalog No. NC9949005
Encompass_Preferred
Platinum-cured peristaltic pump tubing, manufactured with Wacker Silicones HM-6540 material, has the strength and durability normally associated with peroxide-cured tubing.
Catalog No. NC9949005 Supplier Helix Medical Inc Supplier No. 6082523
Platinum-cured peristaltic pump tubing, manufactured with Wacker Silicones HM-6540 material, has the strength and durability normally associated with peroxide-cured tubing.