Learn More
Helix Medical Inc TUBING SIL .040 X .085 50FT
Catalog No. NC9839565
Encompass_Preferred
Platinum-cured peristaltic pump tubing, manufactured with Wacker Silicones HM-6540 material, has the strength and durability normally associated with peroxide-cured tubing. Peristaltic pump tubing undergoes a Special Stabilization Process (SSP), providing the additional benefits of superior strength, performance, and longer life. Tubing offers proven long wear and volumetric consistency over 8-, 12-, and 24-hour process runs.
Catalog No. NC9839565 Supplier Helix Medical Inc Supplier No. 6001105
Platinum-cured peristaltic pump tubing, manufactured with Wacker Silicones HM-6540 material, has the strength and durability normally associated with peroxide-cured tubing. Peristaltic pump tubing undergoes a Special Stabilization Process (SSP), providing the additional benefits of superior strength, performance, and longer life. Tubing offers proven long wear and volumetric consistency over 8-, 12-, and 24-hour process runs.