Learn More
Helix Medical Inc TUBING SIL .058 X .077 50FT
Catalog No. NC9780293
Encompass_Preferred
HelixMark® biomedical/pharmaceutical grade platinum-cured silicone tubing — manufactured with Dow Corning® SILASTIC® Q7-4750 material — provides superior flow properties, is ideal for use in transfer operations as well as other applications requiring nonleachable material, and is able to operate at elevated temperatures while resisting sticking and encrustations. Tubing is packaged double-bagged in crush-resistant boxes with material certification and lot traceability included in every box.
Catalog No. NC9780293 Supplier Helix Medical Inc Supplier No. 6001106
HelixMark® biomedical/pharmaceutical grade platinum-cured silicone tubing — manufactured with Dow Corning® SILASTIC® Q7-4750 material — provides superior flow properties, is ideal for use in transfer operations as well as other applications requiring nonleachable material, and is able to operate at elevated temperatures while resisting sticking and encrustations. Tubing is packaged double-bagged in crush-resistant boxes with material certification and lot traceability included in every box.