Learn More
Helix Medical Inc TUBING SIL .118 X .388 25FT
Catalog No. NC0090771
Encompass_Preferred
Platinum-cured 60 duro peristaltic tubing that provides superior strength, performance and longer life. Meets strength and durability requirements associated with peroxide-cured tubing.
Catalog No. NC0090771 Supplier Helix Medical Inc Supplier No. 60825110
Platinum-cured 60 duro peristaltic tubing that provides superior strength, performance and longer life. Meets strength and durability requirements associated with peroxide-cured tubing.